Merck
  • Identification and characterization of a cis antisense RNA of the parC gene encoding DNA topoisomerase IV of Salmonella enterica serovar Typhi.
Identification and characterization of a cis antisense RNA of the parC gene encoding DNA topoisomerase IV of Salmonella enterica serovar Typhi.
Research in microbiology (2014-05-27)
Isaac Dadzie, Bin Ni, Mingyu Gong, Zhang Ying, Haifang Zhang, Xiumei Sheng, Shungao Xu, Xinxiang Huang
ABSTRACT
Bacterial cis-encoded antisense RNAs are transcribed from the opposite strand of protein coding genes, and their regulatory roles adapt cells to changing environmental conditions. By deep sequencing of the transcriptome of Salmonella enterica serovar Typhi, an antisense RNA that is encoded in cis to the parC gene was found. parC encodes the subunit A component of topoisomerase IV, a class of enzymes that relax both positively and negatively supercoiled DNA and are also required for segregation of daughter chromosomes in bacteria. Transcription of the 871 nucleotide antisense RNA was confirmed by northern blot and RACE analysis to be expressed mostly in the stationary phase of bacterial growth and also upregulated in iron limitation and osmotic stress conditions. Overexpression of the antisense RNA resulted in a significant increase in parC mRNA levels. Further analysis revealed that expression of the antisense RNA stabilizes the target mRNA, probably by protecting it from endoribonucleases. Our findings confirm and add to the ever increasing knowledge of the important role that regulatory antisense RNAs play in bacteria.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)