Merck
  • Photodynamic therapy (PDT) resistance by PARP1 regulation on PDT-induced apoptosis with autophagy in head and neck cancer cells.
Photodynamic therapy (PDT) resistance by PARP1 regulation on PDT-induced apoptosis with autophagy in head and neck cancer cells.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology (2014-06-17)
Jisun Kim, Wonbong Lim, Sangwoo Kim, Sangmi Jeon, Zheng Hui, Kou Ni, Changsu Kim, Yeonggwan Im, Hongran Choi, Okjoon Kim
ABSTRACT
Photodynamic therapy (PDT) is an anticancer treatment that generates excessive reactive oxygen species after photosensitizer treatments following specific wavelength irradiation. In another reports, PDT was regulated with autophagic cell death and apoptotic cell death. However, the mechanism of PDT resistance in PDT-stimulated cell death is unclear. In this study, we determined PDT resistance by autophagy and apoptosis in HP-PDT-treated oral cancer cells. Cells were treated hematoporphyrin and then irradiation with or without inhibitor. Cell lysates were checked protein expression with specific antibody. PDT resistance cells were generated with PDT repeated treatments. In HP-PDT, PDT induced autophagy through mTOR, ATG5, and LC3 in dose-dependent manners. Also, PDT at high dose induced apoptosis through caspase activation and PARP-1. Moreover, PARP-1 inhibitor protected cells against HP-PDT-induced cell death, but not by caspase inhibitor. At low dose of HP, autophagy inhibitor partially protected from HP-PDT-induced cell death. In autophagy phases, at low doses, HP-PDT regulated autophagic cell death through the inhibition of LC3II. Although autophagy inhibitor did not alter cell death directly, autophagy has associated with HP-PDT-induced apoptotic cell death by PARP-1 regulation. Taken together, HP-PDT induces apoptotic cell death with autophagy in oral cancer cells. PDT resistance is related to autophagy by PARP-1 regulation in oral cancer cells.
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)