Merck
  • Cannabidiol exerts sebostatic and antiinflammatory effects on human sebocytes.
Cannabidiol exerts sebostatic and antiinflammatory effects on human sebocytes.
The Journal of clinical investigation (2014-07-26)
Attila Oláh, Balázs I Tóth, István Borbíró, Koji Sugawara, Attila G Szöllõsi, Gabriella Czifra, Balázs Pál, Lídia Ambrus, Jennifer Kloepper, Emanuela Camera, Matteo Ludovici, Mauro Picardo, Thomas Voets, Christos C Zouboulis, Ralf Paus, Tamás Bíró
ABSTRACT
The endocannabinoid system (ECS) regulates multiple physiological processes, including cutaneous cell growth and differentiation. Here, we explored the effects of the major nonpsychotropic phytocannabinoid of Cannabis sativa, (-)-cannabidiol (CBD), on human sebaceous gland function and determined that CBD behaves as a highly effective sebostatic agent. Administration of CBD to cultured human sebocytes and human skin organ culture inhibited the lipogenic actions of various compounds, including arachidonic acid and a combination of linoleic acid and testosterone, and suppressed sebocyte proliferation via the activation of transient receptor potential vanilloid-4 (TRPV4) ion channels. Activation of TRPV4 interfered with the prolipogenic ERK1/2 MAPK pathway and resulted in the downregulation of nuclear receptor interacting protein-1 (NRIP1), which influences glucose and lipid metabolism, thereby inhibiting sebocyte lipogenesis. CBD also exerted complex antiinflammatory actions that were coupled to A2a adenosine receptor-dependent upregulation of tribbles homolog 3 (TRIB3) and inhibition of the NF-κB signaling. Collectively, our findings suggest that, due to the combined lipostatic, antiproliferative, and antiinflammatory effects, CBD has potential as a promising therapeutic agent for the treatment of acne vulgaris.
Product Description
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Adenosine, suitable for cell culture, BioReagent
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine
Adenosine, European Pharmacopoeia (EP) Reference Standard