Merck
  • MiR-210 links hypoxia with cell proliferation regulation in human Laryngocarcinoma cancer.
MiR-210 links hypoxia with cell proliferation regulation in human Laryngocarcinoma cancer.
Journal of cellular biochemistry (2015-02-03)
Jianhong Zuo, Meiling Wen, Mingsheng Lei, Xiang Peng, Xuefeng Yang, Zhigang Liu
ABSTRACT
The microRNA hsa-miR-210 (miR-210) is associated with hypoxia; however its function has not fully identified. In the present study, we aim to detect its role concerning proliferation in Laryngocarcinoma. We found that miR-210 was highly expressed in hypoxia, which inhibited proliferation by inducing cell cycle arrest in G1/G0 as well as apoptosis. We further identified that miR-210 targeted fibroblast growth factor receptor-like 1 (FGFRL1). Down regulation of FGFRL1 decreased cell proliferation by promoting proportion of cells in G1/G0 phase and decreasing in S and G2/M phases. Moreover, overexpression of FGFRL1 effectively released the miR-210-induced suppression of SCC10A cell proliferation. Expression of miR-210 repressed tumor xenograft growth in vivo as well. Together, our findings reveal a new mechanism of adaptation to hypoxia that miR-210 inhibits the proliferation via inducing cell cycle arrest and apoptosis by the targeting of FGFRL1. J. Cell. Biochem. 116: 1039-1049, 2015. © 2015 Wiley Periodicals, Inc.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fgfrl1
Sigma-Aldrich
Methyl Green, zinc chloride salt, Dye content 80 %
Sigma-Aldrich
Methyl Green, zinc chloride salt, certified by the BSC, Dye content 85 %
Sigma-Aldrich
MISSION® esiRNA, targeting human FGFRL1
Sigma-Aldrich
Methyl Green, zinc chloride salt, suitable for microscopy
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Methyl Green, zinc chloride salt, ~85%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)