Merck
  • A family with various symptomatology suggestive of Anderson-Fabry disease and a genetic polymorphism of alpha galactosidase A gene.
A family with various symptomatology suggestive of Anderson-Fabry disease and a genetic polymorphism of alpha galactosidase A gene.
Clinical biochemistry (2014-10-05)
Antonino Tuttolomondo, Giovanni Duro, Rosaria Pecoraro, Irene Simonetta, Salvatore Miceli, Paolo Colomba, Carmela Zizzo, Tiziana Di Chiara, Rosario Scaglione, Vittoriano Della Corte, Francesca Corpora, Antonio Pinto
ABSTRACT
Anderson/Fabry disease expresses a wide range of clinical variability in patients that it is possible to explain referring to a genetic variability with numerous mutations described in the literature (more than 600). We report some clinical cases of some members of a Sicilian family to express phenotypical variability of this disease in subjects with the same genetic mutation The first case was a 59-year-old female. Brain MRI revealed right frontal periventricular white matter of likely vascular-degenerative origin. The proband's alpha galactosidase A activity was 3.7nmol/mL/h. Molecular genetics revealed a polymorphism: -10 C>T; IVS 2-76_80del5; IVS4-16 A>G; IVS6-22 C>T. The second case was a 30year-old male affected by acroparesthesias and hypoidrosis since he was an adolescent. Renal impairment was first detected at age 29; it began with high plasma levels of creatinine and microalbuminuria date. The third case was a 41year-old daughter that presented with acroparesthesias, hypoidrosis since she was very young. The patient's alpha galactosidase A activity was 4.1nmol/mL/h, in whole blood, which is compatible with heterozygote subject for Fabry's disease or healthy control. The fourth case was a male grandson of the proband, 9year-old child. He had a classic gastrointestinal involvement. He complained of recurrent abdominal pain, post prandial bloating and pain. This child's enzyme activity was 1.65nmol/mL/h. In cases 2, 3, and 4, molecular genetics revealed a polymorphism: -10 C>T; IVS 2-76_80del5; IVS4-16 A>G; IVS6-22 C<T DISCUSSION: A recent study reported that IVS4+68 A>G, IVS6-22C>T polymorphisms occurred in 8.9% and 3.7% of the subjects respectively, and the significance of this haplotype in FD pathology remains unknown but possibly suggestive of Anderson/Fabry disease.
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
N-Acetyl-D-galactosamine, ≥98% (HPLC)
Sigma-Aldrich
Hydrochloric acid, BioReagent, 36.5-38.0%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, sterile filtered
Supelco
Water, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – 2-propanol solution, derivatization grade (GC derivatization), ~1.25 M HCl (T), LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Supelco
Water, suitable for ion chromatography
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Water, for TOC analysis
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Water, Deionized