Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Efficacy of RG7787, a next-generation mesothelin-targeted immunotoxin, against triple-negative breast and gastric cancers.
Efficacy of RG7787, a next-generation mesothelin-targeted immunotoxin, against triple-negative breast and gastric cancers.
Molecular cancer therapeutics (2014-09-23)
Christine Alewine, Laiman Xiang, Takao Yamori, Gerhard Niederfellner, Klaus Bosslet, Ira Pastan
ABSTRACT
The RG7787 mesothelin-targeted recombinant immunotoxin (RIT) consists of an antibody fragment targeting mesothelin (MSLN) fused to a 24-kD fragment of Pseudomonas exotoxin A for cell killing. Compared with prior RITs, RG7787 has improved properties for clinical development including decreased nonspecific toxicity and immunogenicity and resistance to degradation by lysosomal proteases. MSLN is a cell surface glycoprotein highly expressed by many solid tumor malignancies. New reports have demonstrated that MSLN is expressed by a significant percentage of triple-negative breast and gastric cancer clinical specimens. Here, panels of triple-negative breast and gastric cancer cell lines were tested for surface MSLN expression, and for sensitivity to RG7787 in vitro and in animal models. RG7787 produced >95% cell killing of the HCC70 and SUM149 breast cancer cell lines in vitro with IC50 < 100 pmol/L. RG7787 was also effective against gastric cancer cell lines MKN28, MKN45, and MKN74 in vitro, with subnanomolar IC50s. In a nude mouse model, RG7787 treatment (2.5 mg/kg i.v. qod ×3-4) resulted in a statistically significant 41% decrease in volumes of HCC70 xenograft tumors (P < 0.0001) and an 18% decrease in MKN28 tumors (P < 0.0001). Pretreatment with paclitaxel (50 mg/kg i.p.) enhanced efficacy, producing 88% and 70% reduction in tumor volumes for HCC70 and MKN28, respectively, a statistically significant improvement over paclitaxel alone (P < 0.0001 for both). RG7787 merits clinical testing for triple-negative breast and gastric cancers.
Product Description
SAFC
L-Glutamine
Sigma-Aldrich
Anti-MSLN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Hematoxylin, certified by the BSC
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Hematoxylin