Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Keratinocyte growth factor-2 is protective in lipopolysaccharide-induced acute lung injury in rats.
Keratinocyte growth factor-2 is protective in lipopolysaccharide-induced acute lung injury in rats.
Respiratory physiology & neurobiology (2014-06-29)
Lin Tong, Jing Bi, Xiaodan Zhu, Guifang Wang, Jie Liu, Linyi Rong, Qin Wang, Nuo Xu, Ming Zhong, Duming Zhu, Yuanlin Song, Chunxue Bai
ABSTRACT
Keratinocyte growth factor-2 (KGF-2) plays a key role in lung development, but its role in acute lung injury has not been well characterized. Lipopolysaccharide instillation caused acute lung injury, which significantly elevated lung wet-to-dry weight ratio, protein and neutrophils in bronchoalveolar lavage fluid (BALF), inhibited surfactant protein A and C expression in lung tissue, and increased pathological injury. Pretreatment with KGF-2 improved the above lung injury parameters, partially restored surfactant protein A and C expression, and KGF-2 given 2-3 days before LPS challenge showed maximum lung injury improvement. Pretreatment with KGF-2 also markedly reduced the levels of TNF-α, MIP-2, IL-1β and IL-6 in BALF and the levels of IL-1β and IL-6 in lung tissue. Histological analysis showed there was increased proliferation of alveolar type II epithelial cells in lung parenchyma, which reached maximal 2 days after KGF-2 instillation. Intratracheal administration of KGF-2 attenuates lung injury induced by LPS, suggesting KGF-2 may be potent in the intervention of acute lung injury.
Sigma-Aldrich
Rat CINC-3 ELISA Kit, for serum, plasma and cell culture supernatant