Merck
  • Synthesis and characterization of Schiff base octaazamacrocyclic complexes and their biological studies.
Synthesis and characterization of Schiff base octaazamacrocyclic complexes and their biological studies.
Journal of photochemistry and photobiology. B, Biology (2014-12-10)
Hina Zafar, Abdul Kareem, Asif Sherwani, Owais Mohammad, Mohammad Azam Ansari, Haris M Khan, Tahir Ali Khan
ABSTRACT
A condensation reaction between 1,2-diphenylethane-1,2-dione dihydrazone (DPEDDH) and dimethyl or diethyloxalate in methanol resulted in a novel Schiff base octaazamacrocyclic ligand, (L): (6,7,14,15-tetraoxa-2,3,10,11-tetraphenyl-1,4,5,8,9,12,13,16-octaazacyclohexadecane-1,3,9,11-tetraene). Subsequently metal complexes of the type [MLX2] and [CuL]X2; (M=Mn(II), Co(II), Ni(II) and Zn(II); X=Cl or NO3) were synthesized by the reaction of the free macrocyclic ligand (L) with the corresponding metal salts in 1:1 molar ratio. These complexes were characterized on the basis of analytical data, molar conductivity and magnetic susceptibility measurements, ESI-mass, IR, NMR ((1)H and (13)C), EPR and electronic spectral studies. The thermal stability of the complexes was also studied by TGA and DTA analyses. These studies show that all the complexes have octahedral arrangement around the metal ions except copper complexes which are square planar. The ligand and its complexes were screened for their antibacterial activity in vitro against Gram-positive (Staphylococcus aureus) and Gram-negative (Escherichia coli) bacteria and were also studied for their anticancer activity against the human cancer cells lines: HeLa (Human cervical carcinoma), MCF7 (Human breast adenocarcinoma) and Hep3B (Human Hepatocellular carcinoma). The recorded IC50 values for the tested compounds show moderate to good cytotoxicity against these cancer cell lines. The copper complex, [CuL]Cl2, showed excellent antimicrobial activity against tested microorganisms which is almost equivalent to the standard drug ciprofloxacin.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Diethyl oxalate, ≥99%
Sigma-Aldrich
Diethyl oxalate, purum, ≥99.0% (GC)
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%