Merck
  • Whole-exome sequencing reveals defective CYP3A4 variants predictive of paclitaxel dose-limiting neuropathy.
Whole-exome sequencing reveals defective CYP3A4 variants predictive of paclitaxel dose-limiting neuropathy.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-11-16)
María Apellániz-Ruiz, Mi-Young Lee, Lara Sánchez-Barroso, Gerardo Gutiérrez-Gutiérrez, Isabel Calvo, Laura García-Estévez, María Sereno, Jesús García-Donás, Beatriz Castelo, Eva Guerra, Luis J Leandro-García, Alberto Cascón, Inger Johansson, Mercedes Robledo, Magnus Ingelman-Sundberg, Cristina Rodríguez-Antona
ABSTRACT
Paclitaxel, a widely used chemotherapeutic drug, can cause peripheral neuropathies leading to dose reductions and treatment suspensions and decreasing the quality of life of patients. It has been suggested that genetic variants altering paclitaxel pharmacokinetics increase neuropathy risk, but the major causes of interindividual differences in susceptibility to paclitaxel toxicity remain unexplained. We carried out a whole-exome sequencing (WES) study to identify genetic susceptibility variants associated with paclitaxel neuropathy. Blood samples from 8 patients with severe paclitaxel-induced peripheral neuropathy were selected for WES. An independent cohort of 228 cancer patients with complete paclitaxel neuropathy data was used for variant screening by DHPLC and association analysis. HEK293 cells were used for heterologous expression and characterization of two novel CYP3A4 enzymes. WES revealed 2 patients with rare CYP3A4 variants, a premature stop codon (CYP3A4*20 allele) and a novel missense variant (CYP3A4*25, p.P389S) causing reduced enzyme expression. Screening for CYP3A4 variants in the independent cohort revealed three additional CYP3A4*20 carriers, and two patients with missense variants exhibiting diminished enzyme activity (CYP3A4*8 and the novel CYP3A4*27 allele, p.L475V). Relative to CYP3A4 wild-type patients, those carrying CYP3A4 defective variants had more severe neuropathy (2- and 1.3-fold higher risk of neuropathy for loss-of-function and missense variants, respectively, P = 0.045) and higher probability of neuropathy-induced paclitaxel treatment modifications (7- and 3-fold higher risk for loss-of-function and missense variants, respectively, P = 5.9 × 10(-5)). This is the first description of a genetic marker associated with paclitaxel treatment modifications caused by neuropathy. CYP3A4 defective variants may provide a basis for paclitaxel treatment individualization.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 5 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 10 mg (per vial)
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate
Fluorescein, European Pharmacopoeia (EP) Reference Standard