Merck
  • Anti-tumor effect of Shu-Gan-Liang-Xue decoction in breast cancer is related to the inhibition of aromatase and steroid sulfatase expression.
Anti-tumor effect of Shu-Gan-Liang-Xue decoction in breast cancer is related to the inhibition of aromatase and steroid sulfatase expression.
Journal of ethnopharmacology (2014-05-09)
Ning Zhou, Shu-Yan Han, Fei Zhou, Ping-ping Li
ABSTRACT
Shu-Gan-Liang-Xue Decoction (SGLXD), a traditional Chinese herbal formula used to ameliorate the hot flushes in breast cancer patients, was reported to have anti-tumor effect on breast cancer. Estrogen plays a critical role in the genesis and evolution of breast cancer. Aromatase and steroid sulfatase (STS) are key estrogen synthesis enzymes that predominantly contribute to the high local hormone concentrations. The present study was to evaluate the anti-tumor effect of SGLXD on estrogen receptor (ER) positive breast cancer cell line ZR-75-1, and to investigate its underlying mechanisms both in vitro and in vivo. The anti-tumor activity of SGLXD in vitro was investigated using the MTT assay. The in vivo anti-tumor effect of SGLXD was evaluated in non-ovariectomized and ovariectomized athymic nude mice. The effect of SGLXD on enzymatic activity of aromatase and STS was examined using the dual-luciferase reporter (DLR) based on bioluminescent measurements. Aromatase and STS protein level were assessed using Western blot assay. SGLXD showed dose-dependent inhibitory effect on the proliferation of ZR-75-1 cells with IC50 value of 3.40 mg/mL. It also suppressed the stimulating effect on cell proliferation of testosterone and estrogen sulfates (E1S). Oral administration of 6 g/kg of SGLXD for 25 days resulted in a reduction in tumor volume in non-ovariectomized and ovariectomized nude mice. The bioluminescent measurements confirmed that SGLXD has a dual-inhibitory effect on the activity of aromatase and STS. Western blot assay demonstrated that the treatment of SGLXD resulted in a decrease in aromatase and STS protein levels both in vitro and in vivo. Our results suggested that SGLXD showed anti-tumor effect on breast cancer cells both in vitro and in vivo. The anti-tumor activity of SGLXD is related to inhibition of aromatase and STS via decreasing their expression. SGLXD may be considered as a novel treatment for ER positive breast cancer.
Sigma-Aldrich
STX64, ≥98% (HPLC)
Sigma-Aldrich
Testosterone, ≥98%
Sigma-Aldrich
Estrone, ≥99%
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Testosterone, VETRANAL®, analytical standard
USP
Estrone, United States Pharmacopeia (USP) Reference Standard
Testosterone, European Pharmacopoeia (EP) Reference Standard
Supelco
Estrone, Pharmaceutical Secondary Standard; Certified Reference Material
Estrone, European Pharmacopoeia (EP) Reference Standard