Merck
  • Global analysis of fungal morphology exposes mechanisms of host cell escape.
Global analysis of fungal morphology exposes mechanisms of host cell escape.
Nature communications (2015-04-01)
Teresa R O'Meara, Amanda O Veri, Troy Ketela, Bo Jiang, Terry Roemer, Leah E Cowen
ABSTRACT
Developmental transitions between single-cell yeast and multicellular filaments underpin virulence of diverse fungal pathogens. For the leading human fungal pathogen Candida albicans, filamentation is thought to be required for immune cell escape via induction of an inflammatory programmed cell death. Here we perform a genome-scale analysis of C. albicans morphogenesis and identify 102 negative morphogenetic regulators and 872 positive regulators, highlighting key roles for ergosterol biosynthesis and N-linked glycosylation. We demonstrate that C. albicans filamentation is not required for escape from host immune cells; instead, macrophage pyroptosis is driven by fungal cell-wall remodelling and exposure of glycosylated proteins in response to the macrophage phagosome. The capacity of killed, previously phagocytized cells to drive macrophage lysis is also observed with the distantly related fungal pathogen Cryptococcus neoformans. This study provides a global view of morphogenetic circuitry governing a key virulence trait, and illuminates a new mechanism by which fungi trigger host cell death.
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Supelco
Fluconazole, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Fluconazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Mannitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Millipore
D-Mannitol, ACS reagent, ≥99.0%, suitable for microbiology
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Fluconazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Terbinafine hydrochloride
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium phosphate dibasic, Vetec, reagent grade, 98%
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%