Merck
  • ES-62 protects against collagen-induced arthritis by resetting interleukin-22 toward resolution of inflammation in the joints.
ES-62 protects against collagen-induced arthritis by resetting interleukin-22 toward resolution of inflammation in the joints.
Arthritis & rheumatology (Hoboken, N.J.) (2014-02-06)
Miguel A Pineda, David T Rodgers, Lamyaa Al-Riyami, William Harnett, Margaret M Harnett
ABSTRACT
The parasitic worm-derived immunomodulator ES-62 protects against disease in the mouse collagen-induced arthritis (CIA) model of rheumatoid arthritis (RA) by suppressing pathogenic interleukin-17 (IL-17) responses. The Th17-associated cytokine IL-22 also appears to have a pathogenic role in autoimmune arthritis, particularly in promoting proinflammatory responses by synovial fibroblasts and osteoclastogenesis. The present study was undertaken to investigate whether the protection against joint damage afforded by ES-62 also reflects suppression of IL-22. The role(s) of IL-22 was assessed by investigating the effects of neutralizing anti-IL-22 antibodies and recombinant IL-22 (rIL-22) on proinflammatory cytokine production, synovial fibroblast responses, and joint damage in mice with CIA in the presence or absence of ES-62. Neutralization of IL-22 during the initiation phase abrogated CIA, while administration of rIL-22 enhanced synovial fibroblast responses and exacerbated joint pathology. In contrast, after disease onset anti-IL-22 did not suppress progression, whereas administration of rIL-22 promoted resolution of inflammation. Consistent with these late antiinflammatory effects, the protection afforded by ES-62 was associated with elevated levels of IL-22 in the serum and joints that reflected a desensitization of the synovial fibroblast responses. Moreover, neutralization of IL-22 during the late effector stage of disease prevented ES-62-mediated desensitization of synovial fibroblast responses and protection against CIA. IL-22 plays a dual role in CIA, being pathogenic during the initiation phase while acting to resolve inflammation and joint damage during established disease. Harnessing of the tissue repair properties of IL-22 by ES-62 highlights the potential for joint-targeted therapeutic modulation of synovial fibroblast responses and consequent protection against bone damage in RA.
Sigma-Aldrich
IL-22 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
IL-22 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
IL-22 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)