Merck
  • The nucleus accumbens shell and the dorsolateral striatum mediate the reinforcing effects of cocaine through a serial connection.
The nucleus accumbens shell and the dorsolateral striatum mediate the reinforcing effects of cocaine through a serial connection.
Behavioural pharmacology (2014-10-18)
Maartje M J Veeneman, Ruth Damsteegt, Louk J M J Vanderschuren
ABSTRACT
The reinforcing and addictive properties of cocaine are thought to rely on the dopaminergic innervation of the striatum. The ventromedial [i.e. nucleus accumbens shell (NAcc) shell] and dorsolateral [dorsolateral striatum (DLS)] regions of the striatum are serially connected, and it is thought that slowly developing neuroadaptations are responsible for the recruitment of the DLS in mediating habitual drug use after extended drug experience. Remarkably, we have recently shown that the DLS is also involved in cocaine self-administration after limited use, to modulate the reinforcing properties of the drug, a function usually ascribed to the NAcc shell. Here, we investigated whether the involvement of the DLS in cocaine reinforcement requires dopaminergic activity within the NAcc shell, by performing a pharmacological disconnection study. We infused the dopamine receptor antagonist α-flupenthixol unilaterally into the NAcc shell and infused this same antagonist into the contralateral DLS, thereby disrupting dopaminergic interconnectivity within the striatum. We show that this disconnection results in increased responding for cocaine under a fixed ratio-1 schedule of reinforcement in rats with limited cocaine experience. These data suggest that a functional dopaminergic interaction between the NAcc shell and the DLS mediates cocaine reinforcement during the early stages of drug use.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
cis-(Z)-Flupenthixol dihydrochloride, ≥98% (HPLC), solid
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Flupentixol dihydrochloride, European Pharmacopoeia (EP) Reference Standard