Merck
  • Kallikrein family proteases KLK6 and KLK7 are potential early detection and diagnostic biomarkers for serous and papillary serous ovarian cancer subtypes.
Kallikrein family proteases KLK6 and KLK7 are potential early detection and diagnostic biomarkers for serous and papillary serous ovarian cancer subtypes.
Journal of ovarian research (2014-12-06)
Ayala Tamir, Ushma Jag, Sreeja Sarojini, Craig Schindewolf, Takemi Tanaka, Rajendra Gharbaran, Hiren Patel, Anil Sood, Wei Hu, Ruzeen Patwa, Patrick Blake, Polina Chirina, Jin Oh Jeong, Heejin Lim, Andre Goy, Andrew Pecora, K Stephen Suh
ABSTRACT
Early detection of ovarian cancer remains a challenge due to widespread metastases and a lack of biomarkers for early-stage disease. This study was conducted to identify relevant biomarkers for both laparoscopic and serum diagnostics in ovarian cancer. Bioinformatics analysis and expression screening in ovarian cancer cell lines were employed. Selected biomarkers were further validated in bio-specimens of diverse cancer types and ovarian cancer subtypes. For non-invasive detection, biomarker proteins were evaluated in serum samples from ovarian cancer patients. Two kallikrein (KLK) serine protease family members (KLK6 and KLK7) were found to be significantly overexpressed relative to normal controls in most of the ovarian cancer cell lines examined. Overexpression of KLK6 and KLK7 mRNA was specific to ovarian cancer, in particular to serous and papillary serous subtypes. In situ hybridization and histopathology further confirmed significantly elevated levels of KLK6 and KLK7 mRNA and proteins in tissue epithelium and a lack of expression in neighboring stroma. Lastly, KLK6 and KLK7 protein levels were significantly elevated in serum samples from serous and papillary serous subtypes in the early stages of ovarian cancer, and therefore could potentially decrease the high "false negative" rates found in the same patients with the common ovarian cancer biomarkers human epididymis protein 4 (HE4) and cancer antigen 125 (CA-125). KLK6 and KLK7 mRNA and protein overexpression is directly associated with early-stage ovarian tumors and can be measured in patient tissue and serum samples. Assays based on KLK6 and KLK7 expression may provide specific and sensitive information for early detection of ovarian cancer.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Formamide, puriss. p.a., ACS reagent, ≥99.5% (GC/T)
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material