Merck
  • CD14 influences host immune responses and alternative activation of macrophages during Schistosoma mansoni infection.
CD14 influences host immune responses and alternative activation of macrophages during Schistosoma mansoni infection.
Infection and immunity (2014-05-29)
Smanla Tundup, Leena Srivastava, Tamas Nagy, Donald Harn
ABSTRACT
Antigen-presenting cell (APC) plasticity is critical for controlling inflammation in metabolic diseases and infections. The roles that pattern recognition receptors (PRRs) play in regulating APC phenotypes are just now being defined. We evaluated the expression of PRRs on APCs in mice infected with the helminth parasite Schistosoma mansoni and observed an upregulation of CD14 expression on macrophages. Schistosome-infected Cd14(-/-) mice showed significantly increased alternative activation of (M2) macrophages in the livers compared to infected wild-type (wt) mice. In addition, splenocytes from infected Cd14(-/-) mice exhibited increased production of CD4(+)-specific interleukin-4 (IL-4), IL-5, and IL-13 and CD4(+)Foxp3(+)IL-10(+) regulatory T cells compared to cells from infected wt mice. S. mansoni-infected Cd14(-/-) mice also presented with smaller liver egg granulomas associated with increased collagen deposition compared to granulomas in infected wt mice. The highest expression of CD14 was found on liver macrophages in infected mice. To determine if the Cd14(-/-) phenotype was in part due to increased M2 macrophages, we adoptively transferred wt macrophages into Cd14(-/-) mice and normalized the M2 and CD4(+) Th cell balance close to that observed in infected wt mice. Finally, we demonstrated that CD14 regulates STAT6 activation, as Cd14(-/-) mice had increased STAT6 activation in vivo, suggesting that lack of CD14 impacts the IL-4Rα-STAT6 pathway, altering macrophage polarization during parasite infection. Collectively, these data identify a previously unrecognized role for CD14 in regulating macrophage plasticity and CD4(+) T cell biasing during helminth infection.
Sigma-Aldrich
Anti-IL4 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Human IL-13 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Rat IL-13 ELISA Kit, for cell and tissue lysates
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluorescein, free acid