Merck
  • The glutathione synthesis inhibitor buthionine sulfoximine synergistically enhanced melphalan activity against preclinical models of multiple myeloma.
The glutathione synthesis inhibitor buthionine sulfoximine synergistically enhanced melphalan activity against preclinical models of multiple myeloma.
Blood cancer journal (2014-07-19)
A Tagde, H Singh, M H Kang, C P Reynolds
ABSTRACT
Melphalan (L-PAM) has been an integral part of multiple myeloma (MM) treatment as a conditioning regimen before stem cell transplant (SCT). After initial response, most treated patients experience relapse with an aggressive phenotype. Increased glutathione (GSH) in MM may mediate resistance to L-PAM. We demonstrated that the GSH synthesis inhibitor buthionine sulfoximine (BSO) synergistically enhanced L-PAM activity (inducing 2-4 logs of cell kill) against nine MM cell lines (also in the presence of marrow stroma or cytokines) and in seven primary MM samples (combination indices <1.0). In MM cell lines, BSO significantly (P<0.05) depleted GSH, increased L-PAM-induced single-strand DNA breaks, mitochondrial depolarization, caspase cleavage and apoptosis. L-PAM depleted GSH, but GSH rapidly recovered in a L-PAM-resistant MM cell line unless also treated with BSO. Treatment with N-acetylcysteine antagonized BSO+L-PAM cytotoxicity without increasing GSH. In human MM xenografted into beige-nude-xid mice, BSO significantly depleted MM intracellular GSH and significantly increased apoptosis compared with L-PAM alone. BSO+L-PAM achieved complete responses (CRs) in three MM xenograft models including maintained CRs >100 days, and significantly increased the median event-free survival relative to L-PAM alone. Combining BSO with L-PAM warrants clinical testing in advanced MM.
Sigma-Aldrich
Sodium thiosulfate, purum p.a., anhydrous, ≥98.0% (RT)
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Melphalan, powder
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
α-Tocopherol, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium thiosulfate, ReagentPlus®, 99%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
Supelco
(±)-α-Tocopherol, analytical standard
Supelco
L-Ascorbic acid, analytical standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Sodium Thiosulfate Solution, 2 g/dL in deionized water