Merck
  • Malondialdehyde-acetaldehyde adducts and anti-malondialdehyde-acetaldehyde antibodies in rheumatoid arthritis.
Malondialdehyde-acetaldehyde adducts and anti-malondialdehyde-acetaldehyde antibodies in rheumatoid arthritis.
Arthritis & rheumatology (Hoboken, N.J.) (2014-11-25)
Geoffrey M Thiele, Michael J Duryee, Daniel R Anderson, Lynell W Klassen, Stephen M Mohring, Kathleen A Young, Dathe Benissan-Messan, Harlan Sayles, Anand Dusad, Carlos D Hunter, Jeremy Sokolove, William H Robinson, James R O'Dell, Anthony P Nicholas, Dean J Tuma, Ted R Mikuls
ABSTRACT
Malondialdehyde-acetaldehyde (MAA) adducts are a product of oxidative stress associated with tolerance loss in several disease states. This study was undertaken to investigate the presence of MAA adducts and circulating anti-MAA antibodies in patients with rheumatoid arthritis (RA). Synovial tissue from patients with RA and patients with osteoarthritis (OA) were examined for the presence of MAA-modified and citrullinated proteins. Anti-MAA antibody isotypes were measured in RA patients (n = 1,720) and healthy controls (n = 80) by enzyme-linked immunosorbent assay. Antigen-specific anti-citrullinated protein antibodies (ACPAs) were measured in RA patients using a multiplex antigen array. Anti-MAA isotype concentrations were compared in a subset of RA patients (n = 80) and matched healthy controls (n = 80). Associations of anti-MAA antibody isotypes with disease characteristics, including ACPA positivity, were examined in all RA patients. Expression of MAA adducts was increased in RA synovial tissue compared to OA synovial tissue, and colocalization with citrullinated proteins was found. Increased levels of anti-MAA antibody isotypes were observed in RA patients compared to controls (P < 0.001). Among RA patients, anti-MAA antibody isotypes were associated with seropositivity for ACPAs and rheumatoid factor (P < 0.001) in addition to select measures of disease activity. Higher anti-MAA antibody concentrations were associated with a greater number of positive antigen-specific ACPA analytes (expressed at high titer) (P < 0.001) and a higher ACPA score (P < 0.001), independent of other covariates. MAA adduct formation is increased in RA and appears to result in robust antibody responses that are strongly associated with ACPAs. These results support speculation that MAA formation may be a cofactor that drives tolerance loss, resulting in the autoimmune responses characteristic of RA.
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Supelco
4,4′-Diaminodiphenylmethane, analytical standard
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, for complexometry, ≥99.0%
Sigma-Aldrich
Methyl acetoacetate, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Methyl acetoacetate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Methyl acetoacetate, Arxada quality, ≥99% (GC)