Merck
  • The rescuable function and mechanism of resveratrol on As₂O₃-induced hERG K⁺ channel deficiency.
The rescuable function and mechanism of resveratrol on As₂O₃-induced hERG K⁺ channel deficiency.
Naunyn-Schmiedeberg's archives of pharmacology (2014-08-12)
Xin Zhao, Kai-Ping Zhang, Ting Huang, Cai-Chuan Yan, Li-Rong Liu, Qi-Lei Zhu, Feng-Feng Guo, Chen Liu, Bao-Xin Li
ABSTRACT
Arsenic trioxide (As2O3) is used to treat acute promyelocytic leukemia. However, the cardiotoxicity of long QT syndrome restricts its clinical application. Previous studies showed that As2O3 can damage the human ether-a-go-go-related gene (hERG) current via disturbing its trafficking to cellular membrane. This study aimed to investigate whether the As2O3-insulted hERG channel can be rescued by resveratrol, a recognized cardioprotective agent. The whole-cell patch clamp technique was used to record the hERG current and action potential duration. Co-immunoprecipitation and Western blot assay were applied to determine the function of hERG-Hsp70/Hsp90 chaperone complexes and the expression alteration of protein-folding-related proteins, respectively. Compared with treatment of As2O3 alone, co-treatment with resveratrol successfully restored the current and surface expression of hERG and obviously shortened action potential duration in guinea pig ventricular myocytes. Further experiments demonstrate that resveratrol relieved As2O3-caused endoplasmic reticulum (ER) stress by restoring the function of hERG-Hsp70/Hsp90 chaperone complexes and downregulating the protein expression of ER chaperone proteins (calnexin and calreticulin) and activating transcription factor 6. In conclusion, resveratrol was able to rescue the trafficking deficiency and relieve the ER stress (ERS). Our findings suggest that resveratrol has a potential effect to alleviate the adverse effect of As2O3 on cardiotoxicity.
Product Description
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Anti-HSP90AA1 (ab2) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)