Merck
  • Motility of select ovarian cancer cell lines: effect of extra-cellular matrix proteins and the involvement of PAK2.
Motility of select ovarian cancer cell lines: effect of extra-cellular matrix proteins and the involvement of PAK2.
International journal of oncology (2014-07-23)
Elizabeth Flate, John R D Stalvey
ABSTRACT
The interaction between tumor cells and extracellular matrix (ECM) proteins influences cell migration and the invasive behavior of cancer cells. In this study, we provide experimental evidence that collagen I and fibronectin affect ovarian cancer cell migration. In vitro wound healing assays and transwell migration assays were used to measure both total wound healing and directionality of individually migrating OV2008 and C13 ovarian cancer cells on glass, collagen I and fibronectin. Involvement of p21‑activated kinase 2 (Pak2) in the motility of these cell lines was investigated using a chemical inhibitor as well as siRNA transfection. Culturing ovarian cancer cells on collagen type I (COLL) increased the migratory ability of OV2008 and C13 cells by increasing the directional migration of cells. In contrast, fibronectin (FN) decreased the migratory ability of OV2008 cells by reducing their ability to migrate directionally. When both cell lines are cultured on COLL and treated with increasing concentrations of a PAK inhibitor (IPA-3), there is a dose-dependent response such that there is a decrease in migration with an increase in inhibitor concentration. Further experiments utilizing PAK2 knockdown via siRNA transfection demonstrated significantly reduced migration of OV2008 cells on COLL as compared to those receiving control siRNA. In conclusion, our results indicate that FN and COLL affect the motility of the selected ovarian cancer cells lines and the effect of COLL is likely mediated, at least in part, by PAK2. A better understanding of the molecular events that contribute to tumor invasion and metastasis is crucial for developing novel treatment strategies to improve the long-term survival of women with ovarian cancer. As PAK2 is involved in motility, it should be further explored as a pro-metastatic gene in ovarian cancer.
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
MISSION® esiRNA, targeting human PAK2
Sigma-Aldrich
HEPES, BioUltra, ≥99.5% (T)
SAFC
HEPES
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
SAFC
HEPES
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pak2
Sigma-Aldrich
MISSION® esiRNA, targeting human PKN2