Merck
  • Curcumin ameliorates epithelial-to-mesenchymal transition of podocytes in vivo and in vitro via regulating caveolin-1.
Curcumin ameliorates epithelial-to-mesenchymal transition of podocytes in vivo and in vitro via regulating caveolin-1.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2014-12-03)
Li-na Sun, Zhi-xin Chen, Xiang-chun Liu, Hai-ying Liu, Guang-ju Guan, Gang Liu
ABSTRACT
Epithelial-mesenchymal transition (EMT) is recognized to play a key role in diabetic nephropathy (DN). Curcumin, the main active component of turmeric extracted from the roots of the Curcuma longa plant, has been reported for its anti-fibrotic effects in kidney fibrosis. The purpose of our study was to investigate the effects of curcumin in reversing epithelial-to-mesenchymal transition (EMT) of podocytes in vivo and in vitro. In vivo streptozotocin (STZ)-induced diabetic rats received vehicle or curcumin, and podocytes were treated with high glucose (HG) in the presence or absence of curcumin in vitro. And we investigated the effect of curcumin on HG-induced phosphorylation of cav-1 on the stability cav-1 and β-catenin using immunoprecipitation and fluorescence microscopy analysis. Curcumin treatment dramatically ameliorated metabolic parameters, renal function, morphological parameters in diabetic rats. We found that HG treatment led to significant down-regulation of p-cadherin and synaptopodin, as well as remarkable up-regulation of α-SMA and FSP-1 in vivo and in vitro. Furthermore, curcumin inhibited HG-induced caveolin-1 (cav-1) Tyr(14) phosphorylation associating with the suppression of stabilization of cav-1 and β-catenin. In summary, these findings suggest that curcumin prevents EMT of podocytes, proteinuria, and kidney injury in DN by suppressing the phosphorylation of cav-1, and increasing stabilization of cav-1 and β-catenin.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Citric acid trisodium salt, ≥98% (GC), anhydrous
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Curcumin, primary reference standard
Supelco
Curcumin, matrix substance for MALDI-MS, ≥99.5% (HPLC)
Supelco
Curcumin, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)