Merck
  • Anticancer and immunoregulatory activity of Gynostemma pentaphyllum polysaccharides in H22 tumor-bearing mice.
Anticancer and immunoregulatory activity of Gynostemma pentaphyllum polysaccharides in H22 tumor-bearing mice.
International journal of biological macromolecules (2014-05-17)
Jun Liu, Lihua Zhang, Yingang Ren, Yanli Gao, Li Kang, Qing Qiao
ABSTRACT
A neutral polysaccharide fraction (CGPP) was extracted from Gynostemma pentaphyllum by water extraction and ethanol precipitation. Gas chromatography (GC) analysis showed that the CGPP was mainly composed of mannose, glucose, arabinose, rhamnose, galactose and glucuronic acid in molar ratios of 2.0:2.2:1.3:2.2:1.2:2.5. The present study aimed at evaluating the antitumor potentials of CGPP on the growth of H22 tumor transplanted in mice and the underlying mechanism. The results showed that CGPP (50 and 200mg/kg) could effectively inhibit the solid tumor growth of H22 hepatocarcinoma transplanted in ICR mice. Besides, the body weight, spleen/thymus indexes and splenocytes proliferation of H22 tumor bearing mice were also improved in CGPP-treated groups. Furthermore, the level of the cytokines, such as IL-2, TNF-α and IFN-γ, as well as the activity of natural killer (NK) cells and cytotoxic T lymphocytes (CTL) in tumor-bearing mice were markedly promoted by CGPP oral administration. In addition, CGPP treatment greatly prolonged the survival period in H22 ascites tumor-bearing mice. Taken together, these findings indicate that CGPP has antitumor activity in vivo at least partly via improving immune responses of host organism, and seems to be a safe and effective supplementary agent for the treatment of hepatocellular carcinoma (HCC).
Product Description
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Trifluoroacetic acid, analytical standard
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture