Merck
  • Nutritional composition and antioxidant properties of the sim fruit (Rhodomyrtus tomentosa).
Nutritional composition and antioxidant properties of the sim fruit (Rhodomyrtus tomentosa).
Food chemistry (2014-08-31)
Thi Ngoc Ha Lai, Christelle André, Hervé Rogez, Eric Mignolet, Thi Bich Thuy Nguyen, Yvan Larondelle
ABSTRACT
In this study, detailed chemical properties of sim (Rhodomyrtus tomentosa (Ait.) Hassk.) fruit including nutritional composition, phenolic content and antioxidant capacity were determined for the first time. A 150g serving of sim fruit contained high levels of dietary fibre (69.94-87.43% of Recommended Daily Intake (RDI)), α-tocopherol (38.90-51.87% RDI), manganese (>100% RDI), and copper (44.44% RDI) but low levels of protein (2.63% RDI), lipid (1.59-3.5% RDI), and sugars (5.65% RDI). The predominant fatty acid in the sim fruit sample was linoleic acid (75.36% of total fatty acids). Interestingly, total phenolics (49.21±0.35mg gallic acid equivalent (GAE)/g dry weight (DW)) were particularly high and resulted in a high antioxidant capacity (431.17±14.56μmol Trolox equivalent (TE)/g DW). These results, together with our recent discovery of high amount of piceatannol, a stilbene with potent biological activities, highlight the potential of sim, an under-utilised plant species from South-East Asia, as a new source of health-promoting compounds including dietary fibres, essential fatty acids, and phenolic compounds.
Product Description
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Potassium acetate, meets USP testing specifications
Sigma-Aldrich
Potassium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium acetate, ≥99.0%
Sigma-Aldrich
Potassium acetate, 99.98% trace metals basis
Sigma-Aldrich
Potassium acetate, 98%
Sigma-Aldrich
Potassium acetate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Potassium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium acetate, puriss., meets analytical specification of Ph. Eur., BP, E261, 99-101%
Sigma-Aldrich
Potassium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium acetate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Potassium acetate solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%