Merck
  • Use of ion-pairing reagent for improving iodine speciation analysis in seaweed by pressure-driven capillary electrophoresis and ultraviolet detection.
Use of ion-pairing reagent for improving iodine speciation analysis in seaweed by pressure-driven capillary electrophoresis and ultraviolet detection.
Journal of chromatography. A (2015-01-13)
Jiannan Sun, Dan Wang, Heyong Cheng, Jinhua Liu, Yuanchao Wang, Zigang Xu
ABSTRACT
This study achieved resolution improvement for iodine speciation in the presence of an ion-pairing reagent by a pressure-driven capillary electrophoresis (CE) system. Addition of 0.01mM tetrabutyl ammonium hydroxide (TBAH) as the ion-pairing reagent into the electrophoretic buffer resulted in the complete separation of four iodine species (I(-), IO3(-), mono-iodothyrosine-MIT and di-iodothyrosine-DIT), because of the electrostatic interaction between TBAH and the negatively charged analytes. A +16kV separation voltage was applied along the separation capillary (50μm i.d., 80cm total and 60cm effective) with the inlet grounded. The detection wavelength was fixed at 210nm, and the pressure-driven flow rate was set at 0.12mLmin(-1) with an injected volume of 2μL. The optimal electrolyte consisted of 2mM borate, 2mM TBAH and 80% methanol with pH adjusted to 8.5. Baseline separation of iodine species was achieved within 7min. The detection limits for I(-), IO3(-), MIT and DIT were 0.052, 0.040, 0.032 and 0.025mgL(-1), respectively. The relative standard deviations of peak heights and areas were all below 3% for 5mgL(-1) and 5% for 1mgL(-1). Application of the proposed method was demonstrated by speciation analysis of iodine in two seaweed samples. The developed method offered satisfactory recoveries in the 91-99% range and good precisions (<5%). Good agreement between the determined values by the proposed CE method and the HPLC-ICP-MS method was also obtained. All results proved its great potential in routine analysis of iodine speciation in environmental, food and biological samples.
Product Description
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Supelco
Potassium Iodide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Supelco
Methanol, analytical standard