Merck
  • Stage of lactation and corresponding diets affect in situ protein degradation by dairy cows.
Stage of lactation and corresponding diets affect in situ protein degradation by dairy cows.
Journal of dairy science (2014-10-06)
I Schadt, D R Mertens, P J Van Soest, G Azzaro, G Licitra
ABSTRACT
The influence of stage of lactation and corresponding diets on rates of protein degradation (kd) is largely unstudied. Study objectives were to measure and compare in situ ruminal kd of crude protein (CP) and estimate rumen CP escape (rumen-undegradable protein; RUP) of selected feeds by cows at 3 stages of lactation fed corresponding diets, and to determine the incubation times needed in an enzymatic in vitro procedure, using 0.2 units of Streptomyces griseus protease per percent of true CP, that predicted in situ RUP. Residue CP was measured after in situ fermentation for 4, 8, 12, 24, 36, 48, and 72 h of 5 protein sources and 3 total mixed rations, which were fed to the in situ cows. Two nonlactating (dry) cows and 2 cows each at 190 (mid) and 90 (peak) days of lactation were used. Each pair of cows was offered free-choice diets that differed in composition to meet their corresponding nutrient requirements. Diets had decreasing proportions of forages and contained (dry matter basis) 11.9, 15.1 and 16.4% CP and 54.3, 40.3 and 35.3% neutral detergent fiber, for dry, mid, and peak TMR (TMR1, TMR2, and TMR3), respectively. Intakes were 10.3, 21.4, and 23.8kg of dry matter/d, respectively. Kinetic CP fractions (extractable, potentially degradable, undegradable, or slowly degradable) were unaffected by treatment. Lag time and kd varied among feeds. The kd was faster for all feeds (0.136/h) when incubated in dry-TMR1 cows compared with mid-TMR2 (0.097/h) or peak-TMR3 (0.098/h) cows, and no differences in lag time were detected. Calculated RUP, using estimated passage rates for each cow based on intake, differed between dry-TMR1 (0.382) and mid-TMR2 (0.559) or peak-TMR3 (0.626) cows, with a tendency for mid-TMR2 to be different from peak-TMR3. Using the average kd and lag time obtained from dry-TMR1 to calculate RUP for mid-TMR2 and peak-TMR3 cows using their passage rates reduced RUP values by 6.3 and 9.5 percentage units, respectively. Except for that of herring meal, in vitro residue CP at 6, 12, and 48h of enzymatic hydrolysis was correlated (r=0.90) with in situ RUP of peak-TMR3, mid-TMR2, and dry-TMR1, respectively. Although confounded within treatments, stage of lactation, diet, and intake appeared to affect CP degradation parameters and RUP. Using kd from nonlactating cows, or the RUP calculated from them, may bias diet evaluation or ration formulation for lactating cows. In addition, enzymatic in vitro predictions of RUP should be measured using incubation times that are appropriate for lactating cows.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Diethylene glycol, ReagentPlus®, 99%
Sigma-Aldrich
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
Sigma-Aldrich
Diethylene glycol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium sulfite, BioUltra, anhydrous, ≥98% (RT)
Sigma-Aldrich
Sodium sulfite, BioXtra, ≥98%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Supelco
Diethylene glycol, analytical standard
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
Diethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Diethylene glycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium sulfite, anhydrous, Redi-Dri, reagent grade, ≥98%
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Sodium sulfite, puriss., meets analytical specification of Ph. Eur., BP, NF, anhydrous, 95.0-100.0% (iodometric)
Sigma-Aldrich
Sodium sulfite, ≥98%
Sigma-Aldrich
Sodium sulfite, ACS reagent, ≥98.0%
Sigma-Aldrich
Sodium sulfite, puriss. p.a., reag. Ph. Eur., anhydrous, 98-100.0% (iodometric)
Supelco
Sodium sulfite, analytical standard
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)