Merck
  • Quinazoline based small molecule exerts potent tumour suppressive properties by inhibiting PI3K/Akt/FoxO3a signalling in experimental colon cancer.
Quinazoline based small molecule exerts potent tumour suppressive properties by inhibiting PI3K/Akt/FoxO3a signalling in experimental colon cancer.
Cancer letters (2015-01-03)
Asif Khurshid Qazi, Aashiq Hussain, Saima Khan, Mushtaq A Aga, Akanksha Behl, Shakir Ali, Shashank Kumar Singh, Subhash Chandra Taneja, Bhahwal Ali Shah, Ajit Kumar Saxena, Dilip Manikrao Mondhe, Abid Hamid
ABSTRACT
Deregulation of PI3K signalling pathway is strongly involved in pathology of cancer and development of resistance in tumour cells. Here, we report that pharmacologically active vasicinone analogue, RLX (7, 8, 9, 10-Tetrahydroazepino [2, 1-b] quinazolin-12-(6H)-on), exhibited potent anticancer activities both in vitro and in vivo. In this study, RLX treatment displayed strong inhibition of proliferation against various cancer cell lines. However, colon cancer cells were found to be the most sensitive towards RLX mediated inhibition of proliferation. The result showed that RLX treatment followed strong concentration dependent inhibition of HCT-116 cell proliferation and colony formation. RLX treatment to HCT-116 was observed to be associated with down-regulation of p110α and p85 subunits of PI3K thereby decreasing the expression of subsequent downstream effector proteins. Interestingly, silencing of PI3K gene by siRNA in combination with RLX confirmed the anti-proliferation effect of RLX against HCT-116 cells and is mediated by the PI3K pathway. We also found that RLX induced sub-G1 arrest and mitochondrial potential loss followed by pFoxO3a(Thr32) nuclear-cytoplasmic translocation inhibition. Moreover, RLX treatment in in vivo models substantially resulted in a tumour growth inhibition. Overall, our findings reveal the functional role of the PI3K/Akt/FoxO3a pathway that gets deregulated in cancer and suggests its simultaneous targeting by RLX thereby further identifying the compound as a potent inhibitor of the PI3K/Akt/FoxO3a pathway under in vitro and tumour regression in vivo.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Adenosine 5′-phosphosulfate sodium salt, ≥85%
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)