Merck
  • Bioisosteric replacement of an acylureido moiety attached to an indolin-2-one scaffold with a malonamido or a 2/4-pyridinoylamido moiety produces a selectively potent Aurora-B inhibitor.
Bioisosteric replacement of an acylureido moiety attached to an indolin-2-one scaffold with a malonamido or a 2/4-pyridinoylamido moiety produces a selectively potent Aurora-B inhibitor.
European journal of medicinal chemistry (2014-07-19)
Hsiao-Chun Wang, Ajit Dhananjay Jagtap, Pei-Teh Chang, Jia-Rong Liu, Chih-Peng Liu, Hsiang-Wen Tseng, Grace Shiahuy Chen, Ji-Wang Chern
ABSTRACT
Bioisosteric replacement of acylureido moiety in 6-acylureido-3-pyrrolylmethylidene-2-oxoindoline derivatives resulted in a series of malonamido derivatives with indolin-2-one scaffold (11-14). Further conformational restrictions of the malonamido moiety led to 2-oxo-1,2-dihydropyridine (21-25) or a 4-oxo-1,4-dihydropyridine derivatives (31-36). 4-Oxo-1,4-dihydropyridine derivatives were more potent Aurora B inhibitors than their 2-oxo-1,2-dihydropyridine counterparts and demonstrated cytotoxicities against A549 and HepG2 cells in the submicromolar range. In A549 cells, 31h decreased phosphorylation of histone H3, triggered polyploidy, induced expression of pro-apoptotic Fas and FasL with subsequent activation of caspase 8, resulting into apoptosis. In a Huh7-xenograft mouse model, 31h demonstrated potent in vivo efficacy with a daily dose of 5 mg/kg.
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
Supelco
1-Methyl-2-pyrrolidinone, analytical standard
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, suitable for HPLC, ≥99%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ACS reagent, ≥99.0%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, biotech. grade, ≥99.7%