Merck
  • In vitro and in silico studies to explore structural features of flavonoids for aldehyde oxidase inhibition.
In vitro and in silico studies to explore structural features of flavonoids for aldehyde oxidase inhibition.
Archiv der Pharmazie (2014-06-24)
Maryam Hamzeh-Mivehroud, Seifullah Rahmani, Mohammad-Ali Hosseinpour Feizi, Siavoush Dastmalchi, Mohammad-Reza Rashidi
ABSTRACT
Aldehyde oxidase (AO), an important enzyme in the biotransformation of drugs and xenobiotics, is inhibited by flavonoids. This enzyme can metabolize both aldehydes and N-heterocycles. In this work, a set of 15 flavonoids was assessed for inhibitory activity on the AO oxidation of vanillin as an aldehyde substrate. Spectrophotometrically determined IC50 values showed that myricetin, quercetin, and epicatechin were the most potent inhibitors. The results also revealed that the inhibition of vanillin oxidation by flavonoids was stronger than that of phenanthridine oxidation (an N-heterocyclic substrate) as reported previously. In order to investigate the important structural features responsible for the inhibitory effects of the studied flavonoids, a quantitative structure-activity relationship (QSAR) analysis was performed. This study showed that the size of the flavonoids was the most important factor inversely affecting their potencies. The QSAR model can be used more broadly to predict the AO inhibitory activity of flavonoid-like structures for application in food-drug and drug-drug interaction studies.
Product Description
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Naringenin, natural (US), 98%
Supelco
Genistein, analytical standard
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%