Merck
  • HBP1 promoter methylation augments the oncogenic β-catenin to correlate with prognosis in NSCLC.
HBP1 promoter methylation augments the oncogenic β-catenin to correlate with prognosis in NSCLC.
Journal of cellular and molecular medicine (2014-06-05)
Ruo-Chia Tseng, Way-Ren Huang, Su-Feng Lin, Pei-Chen Wu, Han-Shui Hsu, Yi-Ching Wang
ABSTRACT
β-catenin nuclear accumulation is frequently identified in human non-small cell lung cancer (NSCLC). The HMG-box transcription factor 1 (HBP1) is a known repressor of β-catenin transactivation. However, the role of HBP1 in relation to β-catenin nuclear accumulation has not been addressed in human cancer patients. In addition, the mechanism of HBP1 gene alteration in NSCLC remains unclear, although HBP1 mutation and gene deletion of HBP1 are reported in breast and colon cancers. Here, we demonstrate that HBP1 acts as a tumour suppressor and serves as a prognostic biomarker in NSCLC clinical and cell models. The immunohistochemistry data indicated that 30.5% (25/82) of tumours from NSCLC patients showed absence or low expression of HBP1 protein. A significant inverse correlation between mRNA/protein expression and promoter hypermethylation suggested that promoter hypermethylation is responsible for low expression of HBP1 in NSCLC patients. Reactivation of HBP1 expression by demethylation reagent or ectopic expression of HBP1 suppressed β-catenin transactivation. Conversely, HBP1 knockdown increased β-catenin transactivation. Importantly, preserved expression of HBP1 had a significantly protective effect on prognosis in patients with β-catenin nuclear accumulation, suggesting that low expression of HBP1 in NSCLC patients with β-catenin nuclear accumulation was one of the major determinants of prognosis. Our data from cellular and clinical models suggest that HBP1 is a suppressor of cancer progression, making it a potential prognostic predictor and therapeutic target to attenuate lung cancer progression.
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
MISSION® esiRNA, targeting human HBP1
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Supelco
Glycerol, analytical standard
SAFC
Tromethamine
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material