Merck
  • Control of homeostatic and pathogenic balance in adipose tissue by ganglioside GM3.
Control of homeostatic and pathogenic balance in adipose tissue by ganglioside GM3.
Glycobiology (2014-10-12)
Masakazu Nagafuku, Takashige Sato, Saya Sato, Kyoko Shimizu, Toshio Taira, Jin-Ichi Inokuchi
ABSTRACT
Ganglioside GM3 (Siaα2-3Galβ1-4Glcβ1-1Cer) has been known to participate in insulin signaling by regulating the association of the insulin receptor in caveolae microdomains (lipid rafts), which is essential for the execution of the complete insulin metabolic signaling in adipocytes. Macrophage-secreted factors including proinflammatory cytokines, tumor necrosis factor-α and interleukin-1β, in adipose tissues have been known to limit the local adipogenesis and induce insulin resistance; however, the interplay between adipocytes and macrophages upon regulation of GM3 expression is not clear. GM3 was virtually absent in primary adipocytes differentiated from macrophage-depleted mesenteric stromal vesicular cells, which accompanies enhancement of insulin signaling and adipogenesis. We found that the expression of GM3 is governed by soluble factors including steady-state levels of proinflammatory cytokines secreted from resident macrophages. The direct involvement of GM3 in insulin signaling is demonstrated by the fact that embryonic fibroblasts obtained from GM3 synthase (GM3S)-deficient mice have increased insulin signaling, when compared with wild-type embryonic fibroblasts, which in turn leads to enhanced adipogeneis. In addition, GM3 expression in primary adipocytes is increased under proinflammatory conditions as well as in adipose tissue of diet-induced obese mice. Moreover, GM3S-deficient mice fed high-fat diets become obese but are resistant to the development of insulin resistance and chronic low-grade inflammatory states. Thus, GM3 functions as a physiological regulatory factor of the balance between homeostatic and pathological states in adipocytes by modulating insulin signaling in lipid rafts.
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Acetic acid, analytical standard
Supelco
Cyclohexane, analytical standard
Supelco
Diethyl ether, analytical standard
Supelco
Methanol, analytical standard
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%