Merck
  • Behaviour of liposomes loaded with bovine serum albumin during in vitro digestion.
Behaviour of liposomes loaded with bovine serum albumin during in vitro digestion.
Food chemistry (2015-01-13)
Weilin Liu, Aiqian Ye, Wei Liu, Chengmei Liu, Jianzhong Han, Harjinder Singh
ABSTRACT
This study examined the stability of liposomes loaded with negatively charged protein (bovine serum albumin, BSA) during in vitro digestion. Zeta-potential and morphology measurements confirmed that BSA-loaded liposomes were successfully prepared, with an encapsulation efficiency of around 34%. The encapsulated BSA and the integrity of the liposomes remained unchanged with time when the liposomes were digested in a simulated gastric environment, suggesting that the liposomal membrane protected the entrapped BSA from pepsin hydrolysis. BSA-loaded liposomes exhibited lower stability in simulated intestinal fluid, as shown by damaged membranes and the release of free fatty acids. Also, lipolysis kinetics revealed that bile salts and ionic strength could facilitate a high level of free fatty acid release. This work further supplemented our knowledge about the effects of gastrointestinal digestion conditions on liposomal properties and provided valuable information for the design of liposome formulations for the food and health care industries.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
(±)-α-Tocopherol, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)