Merck
  • Alkaline phosphatase activity-guided isolation of active compounds and new dammarane-type triterpenes from Cissus quadrangularis hexane extract.
Alkaline phosphatase activity-guided isolation of active compounds and new dammarane-type triterpenes from Cissus quadrangularis hexane extract.
Journal of ethnopharmacology (2014-12-03)
Thanika Pathomwichaiwat, Pannee Ochareon, Noppamas Soonthornchareonnon, Zulfiqar Ali, Ikhlas A Khan, Sompop Prathanturarug
ABSTRACT
The stem of Cissus quadrangularis L. (CQ) is used in traditional medicine to treat bone fractures and swelling. Anti-osteoporotic activity of CQ hexane extract has been reported, but the active compounds in this extract remain unknown. Thus, we aimed to identify the active compounds in CQ hexane extract using bioassay-guided isolation. The CQ hexane extract was fractionated sequentially with benzene, dichloromethane, ethyl acetate, and methanol. The examination of CQ extract and its fractions was guided by bioassays for alkaline phosphatase (ALP) activity during the differentiation of MC3T3-E1 osteoblastic cells. The cells were treated with or without the CQ extract and its fractions for a period of time, and then the stimulatory effect of the alkaline phosphatase enzyme, a bone differentiation marker, was investigated. The compounds obtained were structurally elucidated using spectroscopic techniques and re-evaluated for activity during bone differentiation. A total of 29 compounds were isolated, viz., triterpenes, fatty acid methyl esters, glycerolipids, steroids, phytols, and cerebrosides. Four new dammarane-type triterpenes were isolated for the first time from nature, and this report is the first to identify this group of compounds from the Vitaceae family. Seven compounds, viz., glycerolipids and squalene, stimulated ALP activity at a dose of 10μg/mL. Moreover, the synergistic effect of these compounds on bone formation was demonstrated. This report describes, for the first time, the isolation of active compounds from CQ hexane extract; these active compounds will be useful for the quality control of extracts from this plant used to treat osteoporosis.
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Streptomycin Ready Made Solution, 100 mg/mL in water
Amphotericin B, European Pharmacopoeia (EP) Reference Standard