Merck
  • Expression of vascular endothelial growth factor in human ocular adnexal lymphoma.
Expression of vascular endothelial growth factor in human ocular adnexal lymphoma.
Investigative ophthalmology & visual science (2014-05-16)
Satoshi Kinoshita, Satoru Kase, Ryo Ando, Zhenyu Dong, Junichi Fukuhara, Yoko Dong, Saori Inafuku, Kousuke Noda, Mika Noda, Atsuhiro Kanda, Susumu Ishida
ABSTRACT
To examine the expression of VEGF in extranodal marginal zone B-cell lymphoma (EMZL) and reactive lymphoid hyperplasia (RLH) of human ocular adnexa, and analyze the correlation with the intratumoral microvessel density (MVD). Twenty-two EMZL and 16 RLH tissues were examined in this study. Paraformaldehyde-fixed, paraffin-embedded tissue sections were processed for immunohistochemistry with antibodies against VEGF and CD20. Vascular endothelial growth factor expression was analyzed using the ELISA and RT-PCR in the EMZL tissues. Microvessel density was determined based on the immunoreactivity for anti-CD34 antibody. Vascular endothelial growth factor immunoreactivity was detected in the cytoplasm of lymphoid cells in EMZL and RLH. ELISA and RT-PCR confirmed VEGF protein and mRNA expressions in the EMZL tissue, respectively. Vascular endothelial growth factor-immunopositive rate in B-cells was significantly higher in 12 conjunctival EMZLs than four RLHs (P < 0.01) and 10 orbital EMZLs than 12 RLHs (P < 0.05). The MVD showed a significant positive correlation with the VEGF-immunopositive rate in conjunctival and orbital EMZLs. This study demonstrated increased VEGF expression in human conjunctival and orbital EMZL compared with that in RLH, suggesting that VEGF plays a significant role in the pathogenesis and tumor angiogenesis of ocular adnexal lymphoma.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Human VEGF-A ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Mouse VEGF ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human VEGF R2 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human VEGF R3 ELISA Kit, for serum, plasma, cell culture supernatant, urine
Sigma-Aldrich
Human VEGF ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat VEGF ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat VEGF ELISA Kit, for cell and tissue lysates