Merck
  • Oligosaccharidic fractions derived from Triticum vulgare extract accelerate tissutal repairing processes in in vitro and in vivo models of skin lesions.
Oligosaccharidic fractions derived from Triticum vulgare extract accelerate tissutal repairing processes in in vitro and in vivo models of skin lesions.
Journal of ethnopharmacology (2014-12-03)
Luca Sanguigno, Massimiliano Minale, Ernesto Vannini, Guido Arato, Rodolfo Riccio, Agostino Casapullo, Maria Chiara Monti, Raffaele Riccio, Silvestro Formisano, Gianfranco Di Renzo, Ornella Cuomo
ABSTRACT
Triticum vulgare has been extensively used in traditional medicine thanks to its properties of accelerating tissue repair. The aqueous extract of Triticum vulgare (TVE) is currently an active component used by Farmaceutici Damor in the manufacture of certain pharmaceutical products already marketed in Italy and abroad under the brand name Fitostimoline(®), in the formulation of cream and medicated gauze and is commonly used for the treatment of decubitus ulcers, sores, burns, scarring delays, dystrophic diseases, and, more broadly, in the presence of problems relating to re-epithelialization or tissue regeneration. The active components of Fitostimoline(®)-based products determine a marked acceleration of tissutal repairing processes, stimulate chemotaxis and the fibroblastic maturation, and significantly increase the fibroblastic index, which are crucial points in the repairing processes. The aim of the present paper was to identify and characterize the active fractions of TVE responsible for the pharmacological effect in tissutal repairing processes. Several fractions obtained from TVE by ultrafiltration procedures and HPAE chromatography were tested to measure their growth-enhancing activity on NIH-3T3 fibroblasts. The healing action of the same fractions, prepared as cream formulation, was assessed in rat subjected to two different models of skin lesion, skin scarification and excision. Our results showed a pro-proliferative effect of the fractions ST-98 and K>1000 in NIH-3T3 fibroblasts. Moreover these fractions formulated as cream preparations were effective also in in vivo models of skin lesion. The results of the present study showed that these active fractions of TVE are responsible for its pro-proliferative effect.
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Supelco
2′,4′,6′-Trihydroxyacetophenone monohydrate, matrix substance for MALDI-MS, ≥99.5%
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
2′,4′,6′-Trihydroxyacetophenone monohydrate, 98%
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture