Merck
  • The antihypersensitive and antiinflammatory activities of a benzofuranone derivative in different experimental models in mice: the importance of the protein kinase C pathway.
The antihypersensitive and antiinflammatory activities of a benzofuranone derivative in different experimental models in mice: the importance of the protein kinase C pathway.
Anesthesia and analgesia (2014-07-11)
Juliana Paula de Souza Nunes, Kathryn Ana Bortolini da Silva, Gislaine Francieli da Silva, Nara Lins Meira Quintão, Rogério Corrêa, Valdir Cechinel-Filho, Fátima de Campos-Buzzi, Rivaldo Niero
ABSTRACT
Benzofuranone (BF1) was synthesized and its effects evaluated on mechanical hypersensitivity and paw edema models induced by different agents and on neuropathic pain induced by partial ligation of the sciatic nerve. An attempt was also made to elucidate the mechanism of action. Swiss mice were used for the tests. Hypersensitivity was induced by intraplantar injection of carrageenan, bradykinin (BK), prostaglandin E2 (PGE2), epinephrine, lipopolysaccharide, or complete Freund adjuvant or by using a neuropathic pain model (evaluated with von Frey filament 0.6 g). The antiinflammatory effects were investigated in a paw edema model induced by carrageenan, PGE2, and BK (measured with a plethysmometer). The involvement of protein kinase C (PKC) was investigated through a nociception model induced by phorbol myristate acetate. BF1 inhibited the hypersensitivity and paw edema induced by intraplantar injection of carrageenan, BK, and PGE2 (P < 0.001), and it was effective in reducing the hypersensitivity evoked by complete Freund adjuvant or epinephrine (P < 0.001) but not by lipopolysaccharide (P = 0.2570). BF1 inhibited the licking behavior induced by phorbol myristate acetate (P < 0.001), suggesting involvement of the PKC pathway. A reduction in hypersensitivity of mice submitted to partial ligation of the sciatic nerve (P < 0.001) was observed, with inhibition of neutrophil migration and interleukin-1β production into the spinal cord. BF1 treatment did not interfere with locomotor activity (P = 0.0783) and thermal withdrawal threshold (P = 0.5953), which are important adverse effects of other analgesics. BF1 has dose-dependent antihypersensitive and antiinflammatory effects in both acute and chronic models of pain and inflammation, possibly mediated through interference with the PKC activation pathway. The easy and fast synthesis of this compound, low-cost, low-concentration-requirement, and once-daily-administration drug suggest it as a candidate for future clinical studies.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Chloral hydrate, crystallized, ≥98.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Chloral hydrate
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%