Merck
  • Furan formation during storage and reheating of sterilised vegetable purées.
Furan formation during storage and reheating of sterilised vegetable purées.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2014-12-20)
Stijn Palmers, Tara Grauwet, Carolien Buvé, Lore Van de Vondel, Biniam T Kebede, Marc E Hendrickx, Ann Van Loey
ABSTRACT
To this day, research for furan mitigation has mostly targeted the levels of food production and handling of prepared foods by the consumer. However, part of the furan concentrations found in commercially available food products might originate from chemical deterioration reactions during storage. A range of individual vegetable purées was stored at two different temperatures to investigate the effects of storage on the furan concentrations of shelf-stable, vegetable-based foods. After 5 months of storage at 35°C (temperature-abuse conditions), a general increase in furan concentrations was observed. The furan formation during storage could be reduced by storing the vegetable purées at a refrigerated temperature of 4°C, at which the furan concentrations remained approximately constant for at least 5 months. Following storage, the vegetable purées were briefly reheated to 90°C to simulate the effect of the final preparation step before consumption. Contrary to storage, furan concentrations decreased as a result of evaporative losses. Both refrigerated storage and the reheating step prior to consumption showed the potential of mitigation measures for furan formation in vegetable-based foods (e.g. canned vegetables, ready-to-eat soups, sauces or baby foods). Next to furan, the vegetable purées were analysed for 2- and 3-methylfuran. Tomato was very susceptible to the formation of both alkylated derivatives of furan, as opposed to the other vegetables in this study. Methylfuran concentrations rapidly decreased during storage, which was contrary to the results observed for furan.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Furan-d4, ≥98 atom % D, ≥99% (CP), contains 0.025 wt. % BHT as stabilizer
Sigma-Aldrich
3-Methylfuran, AldrichCPR
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Furan, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Furan, analytical standard
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%