Merck
  • Effects of tobacco smoking during pregnancy on oxidative stress in the umbilical cord and mononuclear blood cells of neonates.
Effects of tobacco smoking during pregnancy on oxidative stress in the umbilical cord and mononuclear blood cells of neonates.
Journal of biomedical science (2014-12-31)
Ednildes de Almeida Olympio Rua, Marcella Leite Porto, Jean Pierre Louzada Ramos, Breno Valentim Nogueira, Silvana S Meyrelles, Elisardo Corral Vasquez, Thiago C Pereira
ABSTRACT
Although cigarette smoke is known to be a complex mixture of over 4000 substances that can lead to damage through active or passive smoking, its mechanisms and biochemical consequences in pregnancy and neonates are not yet fully understood. Therefore, in the present study, we propose to study the impact of smoking during gestation on the viability of blood mononuclear cells (MNC) from umbilical cords of newborns to assess the degree of oxidative stress and cell viability. After childbirth, the cord blood and the umbilical cord were immediately collected in public hospitals in Greater Vitoria, ES, Brazil. Flow cytometry was used to analyze the cord blood followed by biochemical and histological tests to analyze possible changes in the umbilical cord. Pregnant smokers had a reduction of MNC viability from the umbilical cord (10%), an increase in the production of reactive oxygen species (ROS) and an increase in cell apoptosis (~2-fold) compared to pregnant non-smokers. In the umbilical cord, it was observed an increase of advanced oxidation protein products - AOPP (~2.5-fold) and a loss of the typical architecture and disposition of endothelial cells from the umbilical artery. These data suggest that maternal cigarette smoking during pregnancy (even in small amounts) may compromise the viability of MNC cells and damage the umbilical cord structure, possibly by excessive ROS bioavailability.
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, purum p.a., ≥99.0% (AT)
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Supelco
Potassium Iodide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Propidium iodide solution