Merck
  • Effects of the AT1 receptor blocker candesartan on myocardial ischemia/reperfusion in isolated rat hearts.
Effects of the AT1 receptor blocker candesartan on myocardial ischemia/reperfusion in isolated rat hearts.
The heart surgery forum (2014-11-05)
C Murat Songur, Merve Ozenen Songur, Sinan Sabit Kocabeyoglu, Bilgen Basgut
ABSTRACT
We sought to investigate the effects of the angiotension II receptor blocker candesartan on ischemia-reperfusion injury using a cardioplegia arrested isolated rat heart model. Ischemia-reperfusion injury was induced in isolated rat hearts with 40 minutes of global ischemia followed by a 30-minute reperfusion protocol. Throughout the experiment, constant pressure perfusion was achieved using a Langendorff apparatus. Cardioplegic solution alone, and in combination with candesartan, was administered before ischemia and 20 minutes after ischemia. Post-ischemic recovery of contractile function, left ventricular developed pressure, left ventricular end-diastolic pressure and contraction and relaxation rates were evaluated. In the control group, left ventricular developed pressure, rate pressure product, contraction and relaxation rates and coronary flow significantly decreased but coronary resistance increased following reperfusion. With the administration of candesartan alone, parameters did not differ compared to controls. Contractile parameters improved in the group that received candesartan in combination with the cardioplegia compared to the group that received cardioplegia alone; however, the difference between these two groups was insignificant. In this study, the addition of candesartan to a cardioplegic arrest protocol routinely performed during cardiac surgery did not provide a significant advantage in protection against ischemia-reperfusion injury compared with the administration of cardioplegic solution alone.
Product Description
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride hexahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Magnesium chloride hexahydrate, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Magnesium chloride hexahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium chloride hexahydrate, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Magnesium chloride hexahydrate, ACS reagent, 99.0-102.0%
Sigma-Aldrich
Magnesium chloride hexahydrate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E511, 99-101%, ≤0.0001% Al
Sigma-Aldrich
Magnesium chloride hexahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Calcium chloride, Vetec, reagent grade, 96%
Sigma-Aldrich
Magnesium chloride hexahydrate, Vetec, reagent grade, 98%
Sigma-Aldrich
Magnesium chloride hexahydrate, 99.995% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis