Merck
  • Further insights into the composition, source, and toxicity of PAHs in size-resolved particulate matter in a megacity in China.
Further insights into the composition, source, and toxicity of PAHs in size-resolved particulate matter in a megacity in China.
Environmental toxicology and chemistry (2014-11-18)
Guo-Liang Shi, Xiao-Yu Zhou, Su-Yang Jiang, Ying-Ze Tian, Gui-Rong Liu, Yin-Chang Feng, Gang Chen, Yang-Ke-Xin Liang
ABSTRACT
Concentrations of particulate matter with an aerodynamic diameter less than 10 μm (PM10 ) and PM with an aerodynamic diameter less than 2.5 μm (PM2.5 ), and 16 polycyclic aromatic hydrocarbons (PAHs) were measured. The average concentrations of PM10 and PM2.5 reached 209.75 μg/m(3) and 141.87 μg/m(3) , respectively, and those of ΣPAHs were 41.46 ng/m(3) for PM10 and 36.77 ng/m(3) for PM2.5 . The mass ratio concentrations were 219.23 μg/g and 311.01 μg/g in PM10 and PM2.5 , respectively. Three sources and their contributions for PAHs were obtained. For individual input mode, diesel exhaust contributed 46.77% (PM10 ) and 41.12% (PM2.5 ) for mass concentration and 48.69% (PM10 ) and 39.47% (PM2.5 ) for mass ratio concentration; gasoline exhaust contributed 31.02% (PM10 ) and 39.47% (PM2.5 ) for mass concentration and 28.95% (PM10 ) and 36.46% (PM2.5 ) for mass ratio concentration; and coal combustion contributed 22.22% (PM10 ) and 19.41% (PM2.5 ) for mass concentration and 22.36% (PM10 ) and 15.89% (PM2.5 ) for mass ratio concentration. For combined input mode, the same source categories were obtained. Source contributions to PM10 and PM2.5 were diesel exhaust (40.70% and 36.64%, respectively, for mass concentration; 49.19% and 38.47%, respectively, for mass ratio concentration), gasoline exhaust (35.09% and 38.47%, respectively, for mass concentration; 32.50% and 33.43%, respectively, for mass ratio concentration), and coal combustion (24.21% and 24.89%, respectively, for mass concentration; 18.31% and18.17%, respectively, for mass ratio concentration). Source risk assessment showed that vehicle emission was a significant contributor. The findings can help elucidate sources of PAHs and provide evidence supporting further applications of the Unmix model and additional studies about PAHs. Environ Toxicol Chem 2015;34:480-487. © 2014 SETAC.
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Sigma-Aldrich
Anthracene, reagent grade, 97%
Supelco
Anthracene, analytical standard
Supelco
Acenaphthylene, analytical standard
Sigma-Aldrich
Acenaphthylene, 75%
Sigma-Aldrich
Acenaphthylene, 99%
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Supelco
Anthracene, analytical standard
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Supelco
Acenaphthene, analytical standard
Sigma-Aldrich
Acenaphthene, 99%
Supelco
Acenaphthene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland