Merck
  • Enhancement of the antibiotic activity of aminoglycosides by alpha-tocopherol and other cholesterol derivates.
Enhancement of the antibiotic activity of aminoglycosides by alpha-tocopherol and other cholesterol derivates.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2014-12-03)
Jacqueline C Andrade, Maria Flaviana B Morais-Braga, Glaucia M M Guedes, Saulo R Tintino, Maria A Freitas, Irwin R A Menezes, Henrique D M Coutinho
ABSTRACT
Alpha-tocopherol, one of the most abundant isoforms of vitamin E, is a biologically active liposoluble vitamin and potent antioxidant. It occurs naturally in foods of plant and animal origin. Because of its lipophilic character, it can cause perturbations in the bacterial cell membrane, resulting in damage to components essential for the integrity of the membrane, thereby allowing an increase in permeability. This is the first report of the modulatory effect of alpha-tocopherol in multiresistant bacteria. We evaluated alpha-tocopherol against multiresistant strains of Staphylococcus aureus, Pseudomonas aeruginosa and Escherichia coli, and determined if there was any similarity with the mechanism of modulatory action of cholesterol and ergosterol. When combined with aminoglycosides in a microdilution broth assay, alpha-tocopherol possibly acted through a lipophilic action on the cell envelope, modulating more effectively P. aeruginosa and E. coli, when compared with S. aureus.
Product Description
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
(±)-α-Tocopherol, analytical standard
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
USP
Alpha Tocopherol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Ergosterol, ≥75%
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
α-Tocopherol, ≥95.5%
α-Tocopherol, European Pharmacopoeia (EP) Reference Standard
Supelco
Ergosterol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Amikacin, United States Pharmacopeia (USP) Reference Standard
Ergosterol, European Pharmacopoeia (EP) Reference Standard
Amikacin, European Pharmacopoeia (EP) Reference Standard