Merck
  • Chemical modification of cellulosic biopolymer and its use in removal of heavy metal ions from wastewater.
Chemical modification of cellulosic biopolymer and its use in removal of heavy metal ions from wastewater.
International journal of biological macromolecules (2014-04-08)
A S Singha, Ashish Guleria
ABSTRACT
Use of biological macromolecules for wastewater remediation process has become the topic of intense research mostly driven by growing concerns about the depletion of petroleum oil reserves and environmental problems. So in view of technological significance of cellulosic biopolymers in various fields, the present study is an attempt to synthesize cellulosic biopolymers based graft copolymers using free radical polymerization. The resulting cellulosic polymers were characterized by Fourier transform infrared spectroscopy (FT-IR), Scanning electron microscopy (SEM), X-ray diffraction (XRD) and thermogravimetric (TGA) analysis. Furthermore, modified cellulosic biopolymer was used in removal of Cu(2+), Zn(2+), Cd(2+) and Pb(2+) toxic metal ions from wastewater. The effects of pH, contact time, temperature and metal ions concentration were studied in batch mode experiments. Langmuir and Dubinin-Radushkevich (D-R) models were used to show the adsorption isotherm. The maximum monolayer capacity qm calculated using Langmuir isotherm for Cu(2+), Zn(2+), Cd(2+), Pb(2+) metal ions were 1.209, 0.9623, 1.2609 and 1.295mmol/g, respectively. The thermodynamic parameters ΔH° and ΔG° values for metal ions adsorption on modified cellulosic biopolymer showed that adsorption process was spontaneous as well as exothermic in nature.
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
L-Ascorbic acid, analytical standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Copper(II) sulfate, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Copper(II) sulfate, ReagentPlus®, ≥99%
Sigma-Aldrich
Copper(II) sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99-100.5% (based on anhydrous substance)
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard