Merck
Constitutive apoptosis in equine peripheral blood neutrophils in vitro.
Veterinary journal (London, England : 1997) (2014-09-23)
Timothy J Brazil, Padraic M Dixon, Christopher Haslett, Joanna Murray, Bruce C McGorum
ABSTRACT
The aim of this study was to characterise constitutive apoptosis in equine peripheral blood neutrophils, including assessment of factors that potentially modulate neutrophil survival through alteration of the rate of constitutive apoptosis. Cells underwent spontaneous time-dependent constitutive apoptosis when aged in culture for up to 36 h, developing the structural and functional features of apoptosis observed in many cell types, including human neutrophils. Neutrophils undergoing apoptosis also had diminished zymosan activated serum (ZAS)-stimulated chemiluminescence, but maintained responsiveness to phorbol myristate acetate (PMA). The constitutive rate of equine neutrophil apoptosis was promoted by lipopolysaccharide (LPS), tumour necrosis factor α and phagocytosis of opsonised ovine erythrocytes, while it was inhibited by dexamethasone and ZAS (a source of C5a). Formyl-Met-Leu-Phe, leukotriene B4, platelet activating factor and PMA had no demonstrable effect on equine neutrophil apoptosis. There was a difference between equine and human neutrophil apoptosis in response to LPS and the time-dependence of the response to dexamethasone.
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%