Merck
  • The contribution of Gi/o protein to opioid antinociception in an oxaliplatin-induced neuropathy rat model.
The contribution of Gi/o protein to opioid antinociception in an oxaliplatin-induced neuropathy rat model.
Journal of pharmacological sciences (2014-10-28)
Tomoe Kanbara, Atsushi Nakamura, Keiko Takasu, Koichi Ogawa, Masahiro Shibasaki, Tomohisa Mori, Tsutomu Suzuki, Minoru Hasegawa, Gaku Sakaguchi, Toshiyuki Kanemasa
ABSTRACT
Oxaliplatin is a chemotherapeutic agent that induces chronic refractory neuropathy. To determine whether opioids effectively relieve this chronic neuropathy, we investigated the efficacies of morphine, oxycodone, and fentanyl, and the mechanisms underlying opioid antinociception, in oxaliplatin-induced neuropathy in rats. Rats exhibited significant mechanical allodynia following 2 weeks of chronic oxaliplatin administration. Within the range of doses that did not induce sedation and/or muscle rigidity, morphine (3 mg/kg, subcutaneously, s.c.) and oxycodone (0.3-0.56 mg/kg, s.c.) completely reversed oxaliplatin-induced mechanical allodynia, whereas fentanyl (0.017-0.03 mg/kg, s.c.) showed partial antinociception. The antinociception of the optimal doses of morphine and oxycodone were completely inhibited by pertussis toxin (PTX; 0.5 μg/rat, i.c.v.), a Gi/o protein inhibitor, while the partial effect of fentanyl was not affected in the oxaliplatin model. In the [(35)S]-GTPγS binding assay, activation of μ-opioid receptor by fentanyl, but not by morphine or oxycodone, in the mediodorsal thalamus was significantly reduced in oxaliplatin-treated rats. These results indicate that the lower antinociceptive potency of fentanyl in the oxaliplatin model might in part result from the loss of PTX-sensitive Gi/o protein activation, and the degree of Gi/o protein activation might be related to the potency of antinociception by opioids in this model.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Glucose solution, BioUltra, for molecular biology, ~20% in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt, ≥97% (HPLC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture