Merck
  • Responses to various exposure durations of levonorgestrel during early-life stages of fathead minnows (Pimephales promelas).
Responses to various exposure durations of levonorgestrel during early-life stages of fathead minnows (Pimephales promelas).
Aquatic toxicology (Amsterdam, Netherlands) (2015-02-11)
Matthew D Overturf, Duane B Huggett
ABSTRACT
Pharmaceuticals are routinely detected in the environment; and several of these compounds have been extensively researched due to their potential impacts to the endocrine system of aquatic organisms. The negative reproductive consequences of synthetic progestins in teleost species have only recently been investigated. The current study examined different exposure periods that may be most sensitive for levonorgestrel (LNG) in early-life stages of fathead minnow larvae. Larvae were exposed to a single concentration of LNG (125ng/L) for different durations from fertilized egg through 28 days post hatch (dph) with growth and mRNA expression of FSH, 3β-HSD, 20β-HSD, and CYP19a1 measured. Regardless of the duration of exposure, LNG significantly decreased growth in the fathead minnow larvae at day 28. For both 20β-HSD and CYP19a1, mRNA expression was decreased following LNG exposure durations ≥7 days. 3β-HSD and FSH showed similar trends after exposure to LNG with later stages of development exhibiting decreased expression. 20β-HSD and 3β-HSD were the only transcripts to remain down regulated once larvae were moved to clean water after the 7-14dph LNG exposure. This study is the first to investigate the effects of exposure time to a synthetic progestin on developing fish. Future research is needed to understand what impacts these changes have on adult stages of development.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Supelco
D(−)-Norgestrel, analytical standard
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Levonorgestrel for system suitability 2, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%