Merck
  • Structure-dependent binding and activation of perfluorinated compounds on human peroxisome proliferator-activated receptor γ.
Structure-dependent binding and activation of perfluorinated compounds on human peroxisome proliferator-activated receptor γ.
Toxicology and applied pharmacology (2014-07-08)
Lianying Zhang, Xiao-Min Ren, Bin Wan, Liang-Hong Guo
ABSTRACT
Perfluorinated compounds (PFCs) have been shown to disrupt lipid metabolism and even induce cancer in rodents through activation of peroxisome proliferator-activated receptors (PPARs). Lines of evidence showed that PPARα was activated by PFCs. However, the information on the binding interactions between PPARγ and PFCs and subsequent alteration of PPARγ activity is still limited and sometimes inconsistent. In the present study, in vitro binding of 16 PFCs to human PPARγ ligand binding domain (hPPARγ-LBD) and their activity on the receptor in cells were investigated. The results showed that the binding affinity was strongly dependent on their carbon number and functional group. For the eleven perfluorinated carboxylic acids (PFCAs), the binding affinity increased with their carbon number from 4 to 11, and then decreased slightly. The binding affinity of the three perfluorinated sulfonic acids (PFSAs) was stronger than their PFCA counterparts. No binding was detected for the two fluorotelomer alcohols (FTOHs). Circular dichroim spectroscopy showed that PFC binding induced distinctive structural change of the receptor. In dual luciferase reporter assays using transiently transfected Hep G2 cells, PFCs acted as hPPARγ agonists, and their potency correlated with their binding affinity with hPPARγ-LBD. Molecular docking showed that PFCs with different chain length bind with the receptor in different geometry, which may contribute to their differences in binding affinity and transcriptional activity.
Product Description
Supelco
Heptafluorobutyric acid solution, 0.5 M in H2O, LiChropur, suitable for ion pair chromatography
Supelco
Decanoic acid, analytical standard
Sigma-Aldrich
Decanoic acid, ≥98.0%
Sigma-Aldrich
Undecafluorohexanoic acid, ≥97.0% (T)
Sigma-Aldrich
Decanoic acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Decanoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Perfluoroheptanoic acid, ≥97.0%
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Supelco
Pentadecafluorooctanoic acid solution, 100 μg/mL in methanol, analytical standard
Supelco
Pentadecafluorooctanoic acid, analytical standard
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Perfluoroundecanoic acid, 95%
Sigma-Aldrich
Perfluorotetradecanoic acid, 96%
Sigma-Aldrich
Perfluorononanoic acid, 97%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Heptafluorobutyric acid, ≥99.0% (GC)
Supelco
Heptafluorobutyric acid, suitable for ion chromatography, ≥99.5% (GC)
Sigma-Aldrich
Tricosafluorododecanoic acid, 95%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Perfluorodecanoic acid, 97%
Sigma-Aldrich
Heptafluorobutyric acid, 98%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Oleic acid, analytical standard
Supelco
Oleic acid, Selectophore, ≥99%
Oleic acid, European Pharmacopoeia (EP) Reference Standard