Merck
  • Blockade of the T cell immunoglobulin and mucin domain protein 3 pathway exacerbates sepsis-induced immune deviation and immunosuppression.
Blockade of the T cell immunoglobulin and mucin domain protein 3 pathway exacerbates sepsis-induced immune deviation and immunosuppression.
Clinical and experimental immunology (2014-06-20)
Z Zhao, X Jiang, C Kang, Y Xiao, C Hou, J Yu, R Wang, H Xiao, T Zhou, Z Wen, J Feng, G Chen, Y Ma, B Shen, Y Li, G Han
ABSTRACT
Sepsis is a life-threatening condition, but the pathophysiological basis and biomarkers for the monitoring of sepsis and as targets for therapy remain to be determined. We have shown previously that T cell immunoglobulin and mucin domain protein 3 (Tim-3), a negative immune regulator, is involved in the physiopathology of sepsis, but the underlying mechanisms remain unclear. In the present study, we showed that Tim-3 signalling modulated the response patterns of both macrophages and T helper cells in sepsis. Blockade of the Tim-3 pathway exacerbated sepsis-induced proinflammatory macrophage responses and lymphocyte apoptosis during the early phase of sepsis, and enhanced the shift to anti-inflammatory responses for both macrophages and T helper cells during the late phase of sepsis. Tim-3 signalling was found to regulate CD80 and CD86 expression on macrophages both in vivo and in vitro. Co-culture of T cells with Tim-3 knock-down macrophages led to a biased T helper type 2 (Th2) response, partially explaining how Tim-3 signalling shapes inflammation patterns in vivo. Further studies on this pathway might shed new light on the pathogenesis of sepsis and suggest new approaches for intervention.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Rat IL-10 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat IL-10 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human IL-10 R β ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Mouse IL-10 ELISA Kit, for serum, plasma and cell culture supernatant