Merck
  • Investigations into the decomposition of aminoacyl-substituted monosaccharide scaffolds from a drug discovery library.
Investigations into the decomposition of aminoacyl-substituted monosaccharide scaffolds from a drug discovery library.
Organic & biomolecular chemistry (2015-03-05)
Q Q He, N Wimmer, G Verquin, W Meutermans, V Ferro
ABSTRACT
This study investigated the unexpected decomposition and associated intermediates of compound 1, a specific member of a drug discovery library based on a monosaccharide scaffold. LC/MS and NMR spectroscopic analyses indicated that, under acidic conditions, 1 can be converted into the 4-aminogalactoside 2, due to cleavage of the 4-aminobutanoyl side chain. The reaction occurs most likely through an initial intramolecular amino–amide interaction, followed by an N- to O-acyl transfer of the side chain from C-4 to the C-6 position to form an ester intermediate (5), detectable by NMR, and subsequent hydrolysis. Similar decomposition reactions could be induced in selected compounds with similar structures, containing a free hydroxyl group at C-6 and a 4-aminobutanoyl side chain at C-4 of an aminogalactoside. Furthermore, three model compounds were synthesized without a C-6 hydroxyl group and with different length aminoalkanoyl side chains at the C-4 position. The model compounds all decomposed under acidic conditions, but at different rates and much slower when compared with compound 1, suggesting that both the C-6 hydroxyl group and the length of the side chain have an influence on stability.
Product Description
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
Ethyl acetate, analytical standard
Supelco
Hexane, analytical standard
Supelco
Dichloromethane, analytical standard
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%