Merck
  • A general strategy to prepare different types of polysaccharide-graft-poly(aspartic acid) as degradable gene carriers.
A general strategy to prepare different types of polysaccharide-graft-poly(aspartic acid) as degradable gene carriers.
Acta biomaterialia (2014-12-03)
Hai-Qing Song, Xue-Bo Dou, Rui-Quan Li, Bing-Ran Yu, Na-Na Zhao, Fu-Jian Xu
ABSTRACT
Owing to their unique properties such as low cytotoxicity and excellent biocompatibility, poly(aspartic acid) (PAsp) and polysaccharides are good candidates for the development of new biomaterials. In order to construct better gene delivery systems by combining polysaccharides with PAsp, in this work, a general strategy is described for preparing series of polysaccharide-graft-PAsp (including cyclodextrin (CD), dextran (Dex) and chitosan (CS)) gene vectors. Such different polysaccharide-based vectors are compared systematically through a series of experiments including degradability, pDNA condensation capability, cytotoxicity and gene transfection ability. They possess good degradability, which would benefit the release of pDNA from the complexes. They exhibit significantly lower cytotoxicity than the control 'gold-standard' polyethylenimine (PEI, ∼25kDa). More importantly, the gene transfection efficiency of Dex- and CS-based vectors is 12-14-fold higher than CD-based ones. This present study indicates that properly grafting degradable PAsp from polysaccharide backbones is an effective means of producing a new class of degradable biomaterials.
Sigma-Aldrich
Bis(trichloromethyl) carbonate, purum, ≥99.0% (AT)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Triphosgene, reagent grade, 98%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%