Merck
  • Electrospun nanofibers as a potential controlled-release solid dispersion system for poorly water-soluble drugs.
Electrospun nanofibers as a potential controlled-release solid dispersion system for poorly water-soluble drugs.
International journal of pharmaceutics (2015-01-01)
Urve Paaver, Jyrki Heinämäki, Ivo Laidmäe, Andres Lust, Jekaterina Kozlova, Elen Sillaste, Kalle Kirsimäe, Peep Veski, Karin Kogermann
ABSTRACT
Electrospinning was introduced as a novel technique for preparing controlled-release (CR) amorphous solid dispersions (SD) and polymeric nanofibers of a poorly water-soluble drug. Piroxicam (PRX) was used as a low-dose poorly-soluble drug and hydroxypropyl methylcellulose (HPMC) as an amorphous-state stabilising carrier polymer in nanofibers. Raman spectroscopy, X-ray powder diffraction (XPRD), differential scanning calorimetry (DSC) and scanning electron microscopy (SEM) were used in the physical characterisation of the CR-SD nanofibers. Special attention was paid on the effects of a polymer and solvent system on the solid-state properties and physical stability of nanofibers. The average dry diameter of the electrospun CR-SD nanofibers ranged from 400 to 600 nm (SEM). PRX existed in amorphous form in the nanofibers immediately after fabrication and after a short-term (3-month) aging at low temperature (6-8 °C/0% RH) and ambient room temperature (22 °C/0% RH). At higher temperature and humidity (30 °C/85% RH), however, amorphous PRX in the nanofibers tended to slowly recrystallise to PRX form III. The electrospun CR-SD nanofibers exhibited a short lag-time, the absence of initial burst release and zero-order linear CR dissolution kinetics. In conclusion, electrospinning can be used to fabricate supersaturating CR-SD nanofibers of PRX and HPMC, and to stabilise the amorphous state of PRX.
Sigma-Aldrich
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
1,1,1,3,3,3-Hexafluoro-2-propanol, for GC derivatization, LiChropur, ≥99.8%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%