Merck
  • Autoantibodies specific to D4GDI modulate Rho GTPase mediated cytoskeleton remodeling and induce autophagy in T lymphocytes.
Autoantibodies specific to D4GDI modulate Rho GTPase mediated cytoskeleton remodeling and induce autophagy in T lymphocytes.
Journal of autoimmunity (2015-01-28)
Cristiana Barbati, Cristiano Alessandri, Marta Vomero, Rosa Vona, Tania Colasanti, Davide Vacirca, Serena Camerini, Marco Crescenzi, Monica Pendolino, Simona Truglia, Fabrizio Conti, Tina Garofalo, Maurizio Sorice, Marina Pierdominici, Guido Valesini, Walter Malorni, Elena Ortona
ABSTRACT
T lymphocytes from patients with Systemic Lupus Erythematosus (SLE) display multiple abnormalities, including increased cell activation, abnormal cell death by apoptosis and impairment of autophagy pathway. In the present study we report the presence of specific antibodies to D4GDI, a small GTPase family inhibitor, in a significant percentage (46%) of SLE patient sera. We also found a significant association between the presence of these autoantibodies and hematologic manifestations occurring in these patients. Investigating the possible implication of anti-D4GDI autoantibodies in SLE pathogenesis or progression, we found that these antibodies were capable of binding D4GDI expressed at the lymphocyte surface and triggering a series of subcellular events, including Rho GTPase activation. These antibodies were also able to induce autophagy in T cells from both healthy donors and SLE patients, but only those negative to these antibodies. We can conclude that anti-D4GDI autoantibodies could be capable of triggering important responses in T cells such as cytoskeleton remodeling and autophagy pathway and that, in SLE patients, the chronic exposure to these specific autoantibodies could lead to the selection of autophagy-resistant T cell clones contributing to the pathogenesis of the disease.
Sigma-Aldrich
MISSION® esiRNA, targeting human CD3E
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
Formaldehyde solution, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, BioReagent, ≥36.0% in H2O (T)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Triton X-100, laboratory grade
Supelco
Rapamycin, VETRANAL®, analytical standard
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Histidine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard