Merck
  • Fast and versatile fabrication of PMMA microchip electrophoretic devices by laser engraving.
Fast and versatile fabrication of PMMA microchip electrophoretic devices by laser engraving.
Electrophoresis (2014-08-13)
Ellen Flávia Moreira Gabriel, Wendell Karlos Tomazelli Coltro, Carlos D Garcia
ABSTRACT
This paper describes the effects of different modes and engraving parameters on the dimensions of microfluidic structures produced in PMMA using laser engraving. The engraving modes included raster and vector, while the explored engraving parameters included power, speed, frequency, resolution, line-width, and number of passes. Under the optimum conditions, the technique was applied to produce channels suitable for CE separations. Taking advantage of the possibility to cut-through the substrates, the laser was also used to define solution reservoirs (buffer, sample, and waste) and a PDMS-based decoupler. The final device was used to perform the analysis of a model mixture of phenolic compounds within 200 s with baseline resolution.
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Pyrocatechol, ≥99%
Mesalazine impurity C, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
2-Aminophenol, 99%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)